Learn More
Sigma Aldrich Fine Chemicals Biosciences Superoxide dismutase-polye1MG
Catalog No. 502900448
Encompass
Superoxide dismutase from bovine erythrocytes is a copper and zinc superoxide dismutase. Copper and zinc ions are coordinated by active site histidine residues. It has -barrel structure.
Catalog No. 50-290-0448 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S95491MG
Superoxide dismutase from bovine erythrocytes is a copper and zinc superoxide dismutase. Copper and zinc ions are coordinated by active site histidine residues. It has -barrel structure.