Learn More
Sigma Aldrich Fine Chemicals Biosciences Superoxide Dismutase SOD A
Catalog No. 502905530
Encompass
Superoxide dismutase (SOD) is part of a family of metal ion-containing enzymes (SODs) that are found in most organisms from bacteria to mammals. SODs catalyze the dismutation of superoxide (generated by aerobic respiration) to molecular oxygen and peroxide. Superoxide dismutase form the front line of defence against reactive oxygen species (ROS)-mediated injury and harbor anti-inflammatory activities. The Superoxide Dismutase (SOD) Activity Assay Kit provides a simple and sensitive procedure for measuring SOD enzymatic activity in various sample types. SOD activity is determined by measuring the decrease in superoxide anions (generated by the enzyme xanthine oxidase [XO]). Since superoxide anions interact with the provided WST dye yielding color at 450 nm the decrease in the color signal is proportional to SOD inhibition activity. The kit allows the determination of SOD activity in percent inhibition or in units of activity using the provided SOD Enzyme as a standard. The provided
Catalog No. 50-290-5530 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CS00091KT
Superoxide dismutase (SOD) is part of a family of metal ion-containing enzymes (SODs) that are found in most organisms from bacteria to mammals. SODs catalyze the dismutation of superoxide (generated by aerobic respiration) to molecular oxygen and peroxide. Superoxide dismutase form the front line of defence against reactive oxygen species (ROS)-mediated injury and harbor anti-inflammatory activities. The Superoxide Dismutase (SOD) Activity Assay Kit provides a simple and sensitive procedure for measuring SOD enzymatic activity in various sample types. SOD activity is determined by measuring the decrease in superoxide anions (generated by the enzyme xanthine oxidase [XO]). Since superoxide anions interact with the provided WST dye yielding color at 450 nm the decrease in the color signal is proportional to SOD inhibition activity. The kit allows the determination of SOD activity in percent inhibition or in units of activity using the provided SOD Enzyme as a standard. The provided