Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Superpack Pipette Tips

Catalog No. MP0977988H2 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Quantity:
10 x 96/Box
1000/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Quantity
MP0977988H2 Sterile 10 x 96/Box
MP0977987C2 Non-sterile 1000/Box
MP0977987H2 Non-sterile 10 x 96/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP0977988H2 Supplier MP Biomedicals™ Supplier No. 0977988H2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gives superior results, optimal yield, and minimal sample loss compared to inferior pipette tips. Superpack Pipette Tips are available in bulk or in 10 x 96 sterile or non-sterile racks.

Superior pipette tips

  • Autoclavable
  • Recyclable
  • Very low coefficient of friction
  • Naturally hydrophobic
  • Metal-free
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 5 to 300 μL
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Storage Requirements Room Temp
Quantity 10 x 96/Box
Product Type Pipette Tips
Includes Tips, Packs, Case, Rack
Product Line Superpack
Material Plastic
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.