Learn More
FUJIFILM Biosciences SUPERSEP 50UM 7.5% 17WELL XCEL
Catalog No. NC1051662
Encompass_Preferred
*Orders for this supplier are non-cancellable/non-returnable once they are processed.* SuperSep Phos-tag (50?mol/l), 7.5%, 17well, 100¿00¿.6mm
Catalog No. NC1051662 Supplier FUJIFILM Biosciences Supplier No. 19218001
*Orders for this supplier are non-cancellable/non-returnable once they are processed.* SuperSep Phos-tag (50?mol/l), 7.5%, 17well, 100¿00¿.6mm