Learn More
FUJIFILM Biosciences SUPERSEP50UM 12.5% 17WELL XCEL
Catalog No. NC1051659
Encompass_Preferred
*Orders for this supplier are non-cancellable/non-returnable once they are processed.* SuperSep Phos-tag (50umol/l), 12.5%, 17well, 100?100?6.6mm
Catalog No. NC1051659 Supplier FUJIFILM Biosciences Supplier No. 19918011
*Orders for this supplier are non-cancellable/non-returnable once they are processed.* SuperSep Phos-tag (50umol/l), 12.5%, 17well, 100?100?6.6mm