Learn More
Andwin Scientific Cover glasses, Super Slip, rectangular, 50x24 mm, 10x1 oz
SDP
Catalog No. 50949040
Encompass_Preferred
Finally, your automated coverslipper has the right coverslip to allow it to run smoothly. These “Superslip” coverslips are designed to work in automatic coverslip machines and humid environments without sticking or jamming up in the middle of a run. These new superslips combine specialized production procedures with our unique moisture-resistant packaging which brings you superior coverslips.
Catalog No. 50-949-040 Supplier Andwin Scientific Supplier No. 7223651
Finally, your automated coverslipper has the right coverslip to allow it to run smoothly. These “Superslip” coverslips are designed to work in automatic coverslip machines and humid environments without sticking or jamming up in the middle of a run. These new superslips combine specialized production procedures with our unique moisture-resistant packaging which brings you superior coverslips.