Learn More
Labcon North America 1.5mL SuperSpin Micro Centrifuge Tubes, Clear, in Resealable Bags
Catalog No. NC0419739
Encompass_Preferred
Tube, Microcentrifuge; Labcon; SuperSpin; With flat easy open attached snap caps, side graduations, and frosted writing surface; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Ultra high centrifuging to 40,000 x G; Freezable to -80C; Autoclavable; Capacity: 1.5mL; 500 per resealable bag 10 bags per case; Made in USA
Catalog No. NC0419739 Supplier Labcon North America Supplier No. 30168700009
Tube, Microcentrifuge; Labcon; SuperSpin; With flat easy open attached snap caps, side graduations, and frosted writing surface; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Ultra high centrifuging to 40,000 x G; Freezable to -80C; Autoclavable; Capacity: 1.5mL; 500 per resealable bag 10 bags per case; Made in USA