Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Pipe Clay Triangles
GREENER_CHOICESCI_ED

Catalog No. S09563 Shop All Supertek Scientific Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S09563 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S09563 Supplier Supertek™ Supplier No. CH13050/2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pipeclay stems on iron wire.

Specifications

Grade 7 to 12
Length (Metric) 5 cm
Class Size 1 Lab Group
Product Type Pipe Clay Triangle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.