Learn More
Perkin Elmer US LLC Superthane Rinse/Drain Tubing Hookup Kit for ASX-280/560, Oils Applications
Supplier:  Perkin Elmer US LLC N0782045
Encompass_Preferred
Kit Includes: 1/8 X 3/16 PVDF (KYNAR) couplers (qty. 2) 72 in (1.8 m) of 3/16 (4.8 mm) I.D. Superthane tubing 72 in (2.1 m) of 1/8 (3.2 mm) I.D. Superthane tubing
Catalog No. 50-210-5522