Learn More
Across International SuperVac 5.6 cfm Corrosion-Resist 2-Stage Pump
SDP
Catalog No. 502251298
Encompass_Preferred
Ai SuperVac-5C 5.6 cfm commercial grade vacuum pump now come standard with UL certified, this certification is required by many municipalities, and it also provides a higher standard of equipment safety and lifespan. These pumps are also corrosion resistant and 115V/230V 50/60Hz switchable, backed by our 2-year warranty.
This item is not returnable. View return policy
Catalog No. 50-225-1298 Supplier Across International Supplier No. SV5CUL
Ai SuperVac-5C 5.6 cfm commercial grade vacuum pump now come standard with UL certified, this certification is required by many municipalities, and it also provides a higher standard of equipment safety and lifespan. These pumps are also corrosion resistant and 115V/230V 50/60Hz switchable, backed by our 2-year warranty.