Learn More
Medchemexpress LLC HY-123611 5mg Medchemexpress, Supinoxin CAS:888478-45-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1236115MG
Encompass_Preferred
Medchemexpress, HY-123611 5mg Supinoxin CAS:888478-45-3 Supinoxin (RX-5902) is an orally active inhibitor of phosphorylated-p68 RNA helicase (P-p68) and a potent first-in-class anti-cancer agent. Supinoxin interacts with Y593 phosphorylated-p68 and attenuates the nuclear shuttling of β-catenin. Supinoxin induces cell apoptosis and inhibits growth of TNBC cancer cell lines with IC50s ranging from 10 nM to 20 nM[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9965