Learn More
Medchemexpress LLC Suplatast (Tosilate) | 94055-76-2 | 499.64 | 50 MG
SDP
Catalog No. 5000393861
Encompass_Preferred
Suplatast Tosilate (IPD 1151T) is an orally active Th2 cytokine inhibitor that inhibits both IL-4 and IL-5 production from Th2 cells and suppresses IgE synthesis. It is an anti-allergic agent with antiasthmatic, anti-inflammatory, and antifibrotic activity.
  • Orally active Th2 cytokine inhibitor
  • Inhibits IL-4 and IL-5 production
  • Suppresses IgE synthesis
  • Anti-allergic agent
  • Antiasthmatic activity
  • Anti-inflammatory activity
  • Antifibrotic activity
Catalog No. 50-003-93861 Supplier Medchemexpress LLC Supplier No. HY1700250MG
Suplatast Tosilate (IPD 1151T) is an orally active Th2 cytokine inhibitor that inhibits both IL-4 and IL-5 production from Th2 cells and suppresses IgE synthesis. It is an anti-allergic agent with antiasthmatic, anti-inflammatory, and antifibrotic activity.
  • Orally active Th2 cytokine inhibitor
  • Inhibits IL-4 and IL-5 production
  • Suppresses IgE synthesis
  • Anti-allergic agent
  • Antiasthmatic activity
  • Anti-inflammatory activity
  • Antifibrotic activity