Learn More
Medchemexpress LLC Suplatast (Tosilate) | 94055-76-2 | 99.7% | 499.64 | 1 ML
SDP
Catalog No. 5000393862
Encompass_Preferred
Suplatast Tosilate is an orally active Th2 cytokine inhibitor which can inhibit both IL-4 and IL-5 production from Th2 cells and suppress IgE synthesis. It is an anti-allergic agent that also possesses antiasthmatic, anti-inflammatory, and antifibrotic activities.
  • Orally active Th2 cytokine inhibitor
  • Inhibits IL-4 and IL-5 production from Th2 cells
  • Suppresses IgE synthesis
  • Anti-allergic agent
  • Has antiasthmatic activity
  • Has anti-inflammatory activity
  • Has antifibrotic activity
Catalog No. 50-003-93862 Supplier Medchemexpress LLC Supplier No. HY1700210MM/1ML
Suplatast Tosilate is an orally active Th2 cytokine inhibitor which can inhibit both IL-4 and IL-5 production from Th2 cells and suppress IgE synthesis. It is an anti-allergic agent that also possesses antiasthmatic, anti-inflammatory, and antifibrotic activities.
  • Orally active Th2 cytokine inhibitor
  • Inhibits IL-4 and IL-5 production from Th2 cells
  • Suppresses IgE synthesis
  • Anti-allergic agent
  • Has antiasthmatic activity
  • Has anti-inflammatory activity
  • Has antifibrotic activity