Learn More
Apexbio Technology LLC Suplatast Tosylate 94055-76-2 500mg
SDP
Catalog No. 5000089350
Encompass_Preferred
Suplatast Tosylate (CAS 94055-76-2) is a small-molecule modulator that selectively inhibits Th2-mediated cytokine responses It suppresses the production of interleukin-2 (IL-2) interleukin-5 (IL-5) and interleukin-13 (IL-13) without altering interferon-gamma (IFN- ) levels indicating specific action on Th2 pathways while sparing Th1 responses Suplatast Tosylate is used in biomedical research to investigate Th2-driven immunological mechanisms and to explore therapeutic strategies for diseases characterized by Th2 cytokine dysregulation such as allergic and inflammatory disorders
Catalog No. 50-000-89350 Supplier Apexbio Technology LLC Supplier No. B2152500
Suplatast Tosylate (CAS 94055-76-2) is a small-molecule modulator that selectively inhibits Th2-mediated cytokine responses It suppresses the production of interleukin-2 (IL-2) interleukin-5 (IL-5) and interleukin-13 (IL-13) without altering interferon-gamma (IFN- ) levels indicating specific action on Th2 pathways while sparing Th1 responses Suplatast Tosylate is used in biomedical research to investigate Th2-driven immunological mechanisms and to explore therapeutic strategies for diseases characterized by Th2 cytokine dysregulation such as allergic and inflammatory disorders