Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Suplatast Tosylate 98.0+%, TCI America™
SDP

Catalog No. S08751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08751G 1 g
S0875100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08751G Supplier TCI America Supplier No. S08751G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94055-76-2
Molecular Formula C23H33NO7S2
Molecular Weight (g/mol) 499.64
MDL Number MFCD00867604
InChI Key RYVJQEZJUFRANT-UHFFFAOYNA-N
Synonym suplatast tosilate, suplatast tosylate, suplatast, suplatast tosilate inn, ccris 6596, suplatastum tosilas inn-latin, tosilate de suplatast inn-french, tosilato de suplatast inn-spanish, +--2-4-3-ethoxy-2-hydroxypropoxy phenyl carbamoyl ethyldimethylsulfonium p-tosylate
PubChem CID 71773
IUPAC Name (2-{[4-(3-ethoxy-2-hydroxypropoxy)phenyl]carbamoyl}ethyl)dimethylsulfanium 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S([O-])(=O)=O.CCOCC(O)COC1=CC=C(NC(=O)CC[S+](C)C)C=C1

Specifications

Melting Point 87°C
Quantity 1 g
Formula Weight 499.64
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Suplatast Tosylate
RTECSNumber WR7906000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.