Learn More
Cayman Chemical Suplatstosylate 100mg
SDP
Catalog No. 502988084
Encompass_Preferred
An antiallergic agent; inhibits IL-4 and IL-5 production in conalbumin-stimulated D10.G4.1 murine Th2 cells in a concentration-dependent manner; inhibits ovalbumin-induced increases in eosinophil and total cell numbers, as well as IL-4, IL-5, and IL-13, but not IFN-γ, levels in BALF in an ovalbumin-sensitized mouse model of asthma at 100 mg/kg; inhibits ovalbumin-induced increases in ovalbumin-specific IgE in serum and bronchial hyperresponsiveness to methacholine in the ovalbumin-sensitized mouse model of asthma
Catalog No. 50-298-8084 Supplier Cayman Chemical Supplier No. 30316100 MG
An antiallergic agent; inhibits IL-4 and IL-5 production in conalbumin-stimulated D10.G4.1 murine Th2 cells in a concentration-dependent manner; inhibits ovalbumin-induced increases in eosinophil and total cell numbers, as well as IL-4, IL-5, and IL-13, but not IFN-γ, levels in BALF in an ovalbumin-sensitized mouse model of asthma at 100 mg/kg; inhibits ovalbumin-induced increases in ovalbumin-specific IgE in serum and bronchial hyperresponsiveness to methacholine in the ovalbumin-sensitized mouse model of asthma