Learn More
Promocell GmbH PromoCell Supplement Mix / Airway Epithelial Cell Growth Medium
Catalog No. 50306310
Encompass
PromoCell Airway Epithelial Cell Growth Medium is a serum-free medium optimized for the in vitro cultivation of epithelial cells from large air passages.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for equine, porcine, murine, bat and rat airway epithelial cells.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml || Epinephrine | 0.5 μg / ml || Triiodo-L-thyronine | 6.7 ng / ml || Transferrin (recombinant human) | 10 μg / ml || Retinoic Acid | 0.1 ng / ml |
Catalog No. 50-306-310 Supplier Promocell GmbH Supplier No. C39165
PromoCell Airway Epithelial Cell Growth Medium is a serum-free medium optimized for the in vitro cultivation of epithelial cells from large air passages.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for equine, porcine, murine, bat and rat airway epithelial cells.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml || Epinephrine | 0.5 μg / ml || Triiodo-L-thyronine | 6.7 ng / ml || Transferrin (recombinant human) | 10 μg / ml || Retinoic Acid | 0.1 ng / ml |