Learn More
Promocell GmbH PromoCell Supplement Mix / Chondrocyte Growth Medium
Catalog No. 50306353
Encompass
PromoCell Chondrocyte Growth Medium has been developed for the in vitro cultivation of human chondrocytes. The fetal calf serum in the medium is strictly pretested for optimal performance.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for primary porcine and rat chondrocytes.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.1 ml / ml |
Catalog No. 50-306-353 Supplier Promocell GmbH Supplier No. C39635
PromoCell Chondrocyte Growth Medium has been developed for the in vitro cultivation of human chondrocytes. The fetal calf serum in the medium is strictly pretested for optimal performance.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for primary porcine and rat chondrocytes.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.1 ml / ml |