Learn More
Promocell GmbH PromoCell Supplement Mix / Endothelial Cell Growth Medium
Catalog No. 50306315
Encompass
Endothelial Cell Growth Medium is a low-serum (2% V/V) medium optimized for the cultivation of endothelial cells from large blood vessels.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, murine, caprine, rabbit, elephant, dog, and rat endothelial cells from large vessels as well as for various endothelial cell lines.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.02 ml / ml || Endothelial Cell Growth Supplement | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 0.1 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Heparin | 90 μg / ml || Hydrocortisone | 1 μg / ml |Note: For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2.
Catalog No. 50-306-315 Supplier Promocell GmbH Supplier No. C39215
Endothelial Cell Growth Medium is a low-serum (2% V/V) medium optimized for the cultivation of endothelial cells from large blood vessels.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, murine, caprine, rabbit, elephant, dog, and rat endothelial cells from large vessels as well as for various endothelial cell lines.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.02 ml / ml || Endothelial Cell Growth Supplement | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 0.1 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Heparin | 90 μg / ml || Hydrocortisone | 1 μg / ml |Note: For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2.