Learn More
Promocell GmbH PromoCell Supplement Mix / Endothelial Cell Growth Medium 2
Catalog No. 50306316
Encompass_Preferred
Endothelial Cell Growth Medium 2 is a low-serum (2% V/V) medium that lacks Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains Insulin-like Growth Factor (Long R3 IGF) and Vascular Endothelial Growth Factor (VEGF). Because of its effects on cell metabolism, VEGF may interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.02 ml / ml || Epidermal Growth Factor (recombinant human) | 5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 10 ng / ml || Insulin-like Growth Factor (Long R3 IGF) | 20 ng / ml || Vascular Endothelial Growth Factor 165 (recombinant human) | 0.5 ng / ml || Ascorbic Acid | 1 µg / ml || Heparin | 22.5 μg / ml || Hydrocortisone | 0.2 µg / ml |For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2.
Catalog No. 50-306-316 Supplier Promocell GmbH Supplier No. C39216
Endothelial Cell Growth Medium 2 is a low-serum (2% V/V) medium that lacks Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains Insulin-like Growth Factor (Long R3 IGF) and Vascular Endothelial Growth Factor (VEGF). Because of its effects on cell metabolism, VEGF may interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.02 ml / ml || Epidermal Growth Factor (recombinant human) | 5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 10 ng / ml || Insulin-like Growth Factor (Long R3 IGF) | 20 ng / ml || Vascular Endothelial Growth Factor 165 (recombinant human) | 0.5 ng / ml || Ascorbic Acid | 1 µg / ml || Heparin | 22.5 μg / ml || Hydrocortisone | 0.2 µg / ml |For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2.