Learn More
Promocell GmbH PromoCell Supplement Mix / Endothelial Cell Growth Medium MV
Catalog No. 50306319
Encompass
Endothelial Cell Growth Medium MV is a low-serum (5% V/V) medium developed for the in vitro cultivation of endothelial cells from microvascular vessels, the coronary artery, and the aorta.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, murine, rat and dog endothelial cells from microvascular vessels as well as for various endothelial cell lines.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Endothelial Cell Growth Supplement | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Heparin | 90 µg / ml || Hydrocortisone | 1 µg / ml |Note: For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2. Heparin, included in all our other Endothelial Cell Growth Media formulations, may reduce transfection efficiency.
Catalog No. 50-306-319 Supplier Promocell GmbH Supplier No. C39225
Endothelial Cell Growth Medium MV is a low-serum (5% V/V) medium developed for the in vitro cultivation of endothelial cells from microvascular vessels, the coronary artery, and the aorta.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, murine, rat and dog endothelial cells from microvascular vessels as well as for various endothelial cell lines.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Endothelial Cell Growth Supplement | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Heparin | 90 µg / ml || Hydrocortisone | 1 µg / ml |Note: For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2. Heparin, included in all our other Endothelial Cell Growth Media formulations, may reduce transfection efficiency.