Learn More
Promocell GmbH PromoCell Supplement Mix / Fibroblast Growth Medium 2
Catalog No. 50306328
Encompass
PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium and has been developed for the in vitro cultivation of adult Human Fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts, as well as fibroblast cell lines.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.02 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-328 Supplier Promocell GmbH Supplier No. C39325
PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium and has been developed for the in vitro cultivation of adult Human Fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts, as well as fibroblast cell lines.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.02 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |