Learn More
Promocell GmbH PromoCell Supplement Mix / Preadipocyte Growth Medium
Catalog No. 50306338
Encompass
PromoCell Preadipocyte Growth Medium is a low-serum (5% V/V) medium developed for the undifferentiated expansion of human preadipocytes.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine preadipocytes/adipocytes.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.05 ml / ml || Endothelial Cell Growth Supplement | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Hydrocortisone | 1 μg / ml || Heparin | 90 μg / ml |
Catalog No. 50-306-338 Supplier Promocell GmbH Supplier No. C39425
PromoCell Preadipocyte Growth Medium is a low-serum (5% V/V) medium developed for the undifferentiated expansion of human preadipocytes.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine preadipocytes/adipocytes.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.05 ml / ml || Endothelial Cell Growth Supplement | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Hydrocortisone | 1 μg / ml || Heparin | 90 μg / ml |