Learn More
Promocell GmbH PromoCell Supplement Mix / Skeletal Muscle Cell Diff. Medium
Catalog No. 50306334
Encompass
The Skeletal Muscle Cell Differentiation Medium is a defined and animal-component free medium developed to induce the fusion of skeletal muscle cells to myotubes with typical multinucleated syncytia.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine, rat, canine, and rabbit skeletal muscle cells.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:* Differentiation Medium (Ready-to-use): Includes Basal Medium and SupplementMix* SupplementMix: Contains all media supplements premixed in one vial| Media supplement | Final supplement concentration || Insulin (recombinant human) | 10 μg / ml |
Catalog No. 50-306-334 Supplier Promocell GmbH Supplier No. C39366
The Skeletal Muscle Cell Differentiation Medium is a defined and animal-component free medium developed to induce the fusion of skeletal muscle cells to myotubes with typical multinucleated syncytia.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine, rat, canine, and rabbit skeletal muscle cells.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:* Differentiation Medium (Ready-to-use): Includes Basal Medium and SupplementMix* SupplementMix: Contains all media supplements premixed in one vial| Media supplement | Final supplement concentration || Insulin (recombinant human) | 10 μg / ml |