Learn More
Cytiva AcroPak™ 1000 Capsules with Supor Membrane - 0.8/0.45 µm, gamma-irradiated (1/pkg)
Supplier:  Cytiva 12994
Encompass
Fast, Final Filtration and High Throughputs for Liquid Volumes Up To 150 L•Provides higher throughputs and faster flow rates than similarly-sized competitive devices•Ideal for solutions requiring low protein binding•Saves money by increasing throughput with available built-in prefilter•100% integrity testing assures sterile filtrate•0.1 μm Supor membrane version ensures sterile, mycoplasma-free cell culture media•Fusion-welded components eliminate the potential for release of extractables from sealing adhesives•Sterilization by gamma irradiation eliminates potential contamination by EtO residuals This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1698629