Learn More
Promocell GmbH PromoCell Supplement Pack / Adipocyte Nutrition Medium Contains: C-37315, C-31080, C-31005, C-31200,
Catalog No. 50306343
Encompass
PromoCell Adipocyte Nutrition Medium is a low-serum (3% V/V) medium developed for the in vitro cultivation of mature adipocytes.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine preadipocytes/adipocytes.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:* Nutrition Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Nutrition Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.03 ml / ml || d-Biotin | 8 μg / ml || Insulin (recombinant human) | 0.5 μg / ml || Dexamethasone | 400 ng / ml |
Catalog No. 50-306-343 Supplier Promocell GmbH Supplier No. C39439
PromoCell Adipocyte Nutrition Medium is a low-serum (3% V/V) medium developed for the in vitro cultivation of mature adipocytes.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine preadipocytes/adipocytes.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:* Nutrition Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Nutrition Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.03 ml / ml || d-Biotin | 8 μg / ml || Insulin (recombinant human) | 0.5 μg / ml || Dexamethasone | 400 ng / ml |