Learn More
Promocell GmbH PromoCell Supplement Pack / Endothelial Cell Growth Medium 2 Contains: C-37320, C-30224, C-31063, C-30260, C-30321, C-31700, C-31750, C-31650
Catalog No. 50306314
Encompass
Endothelial Cell Growth Medium 2 is a low-serum (2% V/V) medium that lacks Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains Insulin-like Growth Factor (Long R3 IGF) and Vascular Endothelial Growth Factor (VEGF). Because of its effects on cell metabolism, VEGF may interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.02 ml / ml || Epidermal Growth Factor (recombinant human) | 5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 10 ng / ml || Insulin-like Growth Factor (Long R3 IGF) | 20 ng / ml || Vascular Endothelial Growth Factor 165 (recombinant human) | 0.5 ng / ml || Ascorbic Acid | 1 µg / ml || Heparin | 22.5 μg / ml || Hydrocortisone | 0.2 µg / ml |For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2.
Catalog No. 50-306-314 Supplier Promocell GmbH Supplier No. C39211
Endothelial Cell Growth Medium 2 is a low-serum (2% V/V) medium that lacks Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains Insulin-like Growth Factor (Long R3 IGF) and Vascular Endothelial Growth Factor (VEGF). Because of its effects on cell metabolism, VEGF may interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.02 ml / ml || Epidermal Growth Factor (recombinant human) | 5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 10 ng / ml || Insulin-like Growth Factor (Long R3 IGF) | 20 ng / ml || Vascular Endothelial Growth Factor 165 (recombinant human) | 0.5 ng / ml || Ascorbic Acid | 1 µg / ml || Heparin | 22.5 μg / ml || Hydrocortisone | 0.2 µg / ml |For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2.