Learn More
Promocell GmbH PromoCell Supplement Pack / Fibroblast Growth Medium 3 Contains: C-37340, C-30310, C-31010
Catalog No. 50306330
Encompass
PromoCell Fibroblast Growth Medium 3 has been developed for the in vitro cultivation of adult Human Cardiac Fibroblasts (HCF).Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine cardiac fibroblasts.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.1 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-330 Supplier Promocell GmbH Supplier No. C39350
PromoCell Fibroblast Growth Medium 3 has been developed for the in vitro cultivation of adult Human Cardiac Fibroblasts (HCF).Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine cardiac fibroblasts.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.1 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |