Learn More
Promocell GmbH PromoCell Supplement Pack / Mammary Epithelial Cell Growth Medium Contains: C-30021, C-30226, C-31010, C-31060,
Catalog No. 50306307
Encompass
The PromoCell Mammary Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the mammary gland.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for rat and murine mammary epithelial cells as well as mamma carcinoma cell lines.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml |
Catalog No. 50-306-307 Supplier Promocell GmbH Supplier No. C39110
The PromoCell Mammary Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the mammary gland.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for rat and murine mammary epithelial cells as well as mamma carcinoma cell lines.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml |