Learn More
Promocell GmbH PromoCell Supplement Pack/ Myocyte Growth Medium Contains: C-30222, C-31010, C-37310, C-30320
Catalog No. 50306323
Encompass
The PromoCell Myocyte Growth Medium is a low serum (5% V/V) medium supplemented with well chosen hormones and growth factors. It was developed for the optimal expansion and maintenance of primary cardiac myocytes in culture.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine and primary rat cardiac myocytes.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 0.5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 2 ng / ml || Insulin (recombinant human) | 5 µg / ml |
Catalog No. 50-306-323 Supplier Promocell GmbH Supplier No. C39270
The PromoCell Myocyte Growth Medium is a low serum (5% V/V) medium supplemented with well chosen hormones and growth factors. It was developed for the optimal expansion and maintenance of primary cardiac myocytes in culture.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine and primary rat cardiac myocytes.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 0.5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 2 ng / ml || Insulin (recombinant human) | 5 µg / ml |