Learn More
Promocell GmbH PromoCell Supplement Pack / Skeletal Muscle Cell Growth Medium Contains: C-37310, C-30226, C-30310, C-30700, C-31020, C-31200
Catalog No. 50306331
Encompass
The PromoCell Skeletal Muscle Cell Growth Medium is a low-serum medium (5% V/V) developed for the expansion of human skeletal muscle cell cultures.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine, rat, canine, and rabbit skeletal muscle cells.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Fetuin (bovine) | 50 μg / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 10 μg / ml || Dexamethasone | 0.4 μg / ml |
Catalog No. 50-306-331 Supplier Promocell GmbH Supplier No. C39360
The PromoCell Skeletal Muscle Cell Growth Medium is a low-serum medium (5% V/V) developed for the expansion of human skeletal muscle cell cultures.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine, rat, canine, and rabbit skeletal muscle cells.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Fetuin (bovine) | 50 μg / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 10 μg / ml || Dexamethasone | 0.4 μg / ml |