Learn More
Thermo Scientific™ Supplemented Brucella Broth for Anaerobes
Catalog No. YT3450 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
Catalog No. Quantity
YT3450 10 mL
1 options
This item is not returnable. View return policy
Catalog No. YT3450 Supplier Thermo Scientific™ Supplier No. YT3450
Add to Cart
Add to Cart
This item is not returnable. View return policy
Perform susceptibility testing with a full range of supplies, including this broth, to enhance your Thermo Scientific™ Sensititre™ test program.
Perform high-quality susceptibility testing with a complete range of supplies, including the Thermo Scientific™ Supplemented Brucella Broth for Anaerobes, to complement and enhance your Thermo Scientific™ Sensititre™ test program.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description Supplemented Brucella Broth for Anaerobes
Product Type Microbiology Media
Target Organism Class Anaerobes
Test Type Microbial Susceptibility
No. per Pack 10
Packaging Type 10 mL/Bottle
Quantity 10 mL