Learn More
3M™ Supplied-Air Respirator Air-Control Devices
Catalog No. 19038984 Shop All 3M Products
Change view
Click to view available options
Description:
Air Regulating Valve Assembly
Vortex Cooling Assembly
For Use With (Equipment):
L series loose fitting headgear, whitecap™ helmets and h series hoods, 3M™ belt mounted supplied air respirator
Includes:
Cooling tube with quick-disconnect fitting, waist belt and holder
Valve and waist belt
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description For Use With (Equipment) Includes
19-038-984 Air Regulating Valve Assembly L series loose fitting headgear, whitecap™ helmets and h series hoods, 3M™ belt mounted supplied air respirator Valve and waist belt
19-038-970 Vortex Cooling Assembly L series loose fitting headgear, whitecap™ helmets and h series hoods, 3M™ belt mounted supplied air respirator Cooling tube with quick-disconnect fitting, waist belt and holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19-038-984 Supplier 3M™ Supplier No. 7100331615
Add to Cart
Add to Cart
This item is not returnable. View return policy
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Air-control devices provide positive pressure and regulate the flow and temperature of compressed air.
  • All heating, cooling, and connector assemblies include waist belt and tube with quick-disconnect fitting
  • 3M Catalog# V-100 , 3M Catalog# V-300 , 3M Catalog# V-400
TRUSTED_SUSTAINABILITY
Specifications
Description Air Regulating Valve Assembly
Product Type Flow Control Valve
For Use With (Application) Blasting, assembly, mechanical, painting, metal repair, cleaning
For Use With (Equipment) L series loose fitting headgear, whitecap™ helmets and h series hoods, 3M™ belt mounted supplied air respirator
Includes Valve and waist belt
Model V-300
Pressure 25 to 55 psi