Learn More
Chemglass Life Sciences Filtration Support Assembly, 47MM Fritted, Complete
SDP
Supplier:  Chemglass Life Sciences CG142001
Encompass_Preferred
Filtration Support Assembly, 47MM Fritted, Complete. Glassware system for vacuum filtration using 47mm membranes. Assembly is designed for use with vacuum manifolds or optional plastic coated filtering flask. Support base has a sealed-in coarse porosity fritted disc. Complete assembly includes a 300mL graduated funnel, aluminum clamp, support base and silicone stopper.
Catalog No. CG142001