Learn More
Chemglass Life Sciences Fritted support base for 47 mm membrane filtration assemblies - 40/35 standard taper (base only)
SDP
Supplier:  Chemglass Life Sciences CG142402
Encompass_Preferred
This 47 mm fritted support base is a glass vacuum filtration component designed to hold 47 mm membranes and connect directly to standard taper filtering flasks for solvent and particulate filtration.
  • Supports 47 mm membrane filters.
  • Sealed-in coarse porosity fritted disc for particle retention.
  • Standard taper 40/35 joint for direct connection to filtering flasks.
  • Vacuum port positioned above the drip tip to prevent filtrate entering the vacuum line.
  • Suitable for filtering corrosive solutions, removing particulates from HPLC solvents, and filtration analysis with SPE disks.
Catalog No. NC1841216