Learn More
Wilmad Labglass SUPPORT BASE 90MM 40/35
Supplier:  Wilmad Labglass BP1826090
Encompass_Preferred
Support Base, Fritted Glass. Filter Size: 90mm; Joint Size: [ST] 40/35 outer. Replacement membrane filter Support base for BP-1825-000, BP-1825-002 and BP-1825-004. Supplied with coarse porosity glass frit.
Catalog No. 50-805-058
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov