Learn More
Chemglass Life Sciences Support Base, Rectangular
SDP
Catalog No. 501311207
Encompass_Preferred
Support Base, Rectangular. Base is made of cast iron with a black baked finish. Dimensions are 140 x 200mm. Base has two holes, one centered and one at the end of the short side. Rod is 10mm O.D. x 600mm (24) in height. Total assembled weight is 1Kg.
Catalog No. 50-131-1207 Supplier Chemglass Life Sciences Supplier No. CG3045104
Support Base, Rectangular. Base is made of cast iron with a black baked finish. Dimensions are 140 x 200mm. Base has two holes, one centered and one at the end of the short side. Rod is 10mm O.D. x 600mm (24) in height. Total assembled weight is 1Kg.