Learn More
Hach Company Stand, Support, 178 x 330 mm Base
Catalog No. 501994252 Shop All Hach Company Products
Encompass
Cast iron, white enamel base provides favorable background for observing end points. Also acts as a steady support for buret. Base is 178 x 330 mm (7 x 13") furnished with a 13 x 610 mm (0.5 x 24") aluminum rod with a threaded nut.
Catalog No. 50-199-4252 Supplier Hach Company Supplier No. 32900
Cast iron, white enamel base provides favorable background for observing end points. Also acts as a steady support for buret. Base is 178 x 330 mm (7 x 13") furnished with a 13 x 610 mm (0.5 x 24") aluminum rod with a threaded nut.