Learn More
Chemglass Life Sciences Support, Flask, Tripod, Large, 6" Ring ID X 11.5" Height, PTFE Coated
SDP
Catalog No. 50872561
Encompass_Preferred
Support, Flask, Tripod, Large, 6" Ring ID X 11.5" Height, PTFE Coated. Support designed to hold flask and jacketed reactors. Made of PTFE coated S.S. rod, the upper ring is wrapped in silicone to prevent metal to glass contact. Front portion of ring is cutaway to allow flask having a lower drain valve to be placed in the ring without lifting it over the ring.
Catalog No. 50-872-561 Supplier Chemglass Life Sciences Supplier No. CG304930
Support, Flask, Tripod, Large, 6" Ring ID X 11.5" Height, PTFE Coated. Support designed to hold flask and jacketed reactors. Made of PTFE coated S.S. rod, the upper ring is wrapped in silicone to prevent metal to glass contact. Front portion of ring is cutaway to allow flask having a lower drain valve to be placed in the ring without lifting it over the ring.