Learn More
Chemglass Life Sciences Support/Holding Rod for PT Sensor - Hotplate Probe Support
SDP
Catalog No. NC2385832
Encompass_Preferred
Support holding rod used to secure a PT (temperature) sensor or probe to the vertical support rod of a hotplate. Provides a stable mounting point using a boss head and is sold individually as part of a machined product line.
  • Secures a PT temperature sensor or probe to a vertical support rod on a hotplate.
  • Designed for use with a boss head for reliable clamping.
  • Provides a stable mounting point to maintain probe position during heating or stirring.
  • Sold individually.
  • Part of a machined product line for precision fit.
Catalog No. NC2385832 Supplier Chemglass Life Sciences Supplier No. CG1995V27
Support holding rod used to secure a PT (temperature) sensor or probe to the vertical support rod of a hotplate. Provides a stable mounting point using a boss head and is sold individually as part of a machined product line.
  • Secures a PT temperature sensor or probe to a vertical support rod on a hotplate.
  • Designed for use with a boss head for reliable clamping.
  • Provides a stable mounting point to maintain probe position during heating or stirring.
  • Sold individually.
  • Part of a machined product line for precision fit.