Learn More
BRANSON ULTRASONICS CORPORATION (3P) SUPPORT RACK FOR FS220
Catalog No. NC3721424
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Support Racks, For Model 8800, For Use With: For Model 8800, Support rack, Height: 20 in., 50.8cm, Width: 15 in.
Catalog No. NC3721424 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPN916043
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Support Racks, For Model 8800, For Use With: For Model 8800, Support rack, Height: 20 in., 50.8cm, Width: 15 in.