Learn More
BRANSON ULTRASONICS CORPORATION (3P) SUPPORT RACK FOR FS60
Catalog No. NC3721422
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Support Racks, For Model 3800, For Use With: For Model 3800, Support rack, Height: 12 in., 30.48cm, Width: 8 in.
Catalog No. NC3721422 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPN916041
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Support Racks, For Model 3800, For Use With: For Model 3800, Support rack, Height: 12 in., 30.48cm, Width: 8 in.