Learn More
Chemglass Life Sciences Support Ring with Opening, Large, 85mm ID, 190mm Support Rod Length
SDP
Catalog No. 501311134
Encompass_Preferred
Support Ring with Opening, Large, 85mm ID, 190mm Support Rod Length. Manufactured from steel wire and zinc plated, rings have an opening to permit items such as separatory funnels to be inserted without removing the support ring from the base. Available in three sizes.
Catalog No. 50-131-1134 Supplier Chemglass Life Sciences Supplier No. CG1948L12
Support Ring with Opening, Large, 85mm ID, 190mm Support Rod Length. Manufactured from steel wire and zinc plated, rings have an opening to permit items such as separatory funnels to be inserted without removing the support ring from the base. Available in three sizes.