Learn More
Hach Company Stand, Buret Support, 127 x 203 mm Base
Catalog No. 501994391 Shop All Hach Company Products
Encompass
Heavy black enamel, cast iron base and zinc-plated rod. Base is 127 x 203 mm (5 x 8") with an 8 x 508 mm (0.3 x 20") rod.
Catalog No. 50-199-4391 Supplier Hach Company Supplier No. 56300
Heavy black enamel, cast iron base and zinc-plated rod. Base is 127 x 203 mm (5 x 8") with an 8 x 508 mm (0.3 x 20") rod.