Learn More
Chemglass Life Sciences Support Ring with Clamp, Large, 85mm ID, 200mm OAL
SDP
Catalog No. 501311213
Encompass_Preferred
Support Ring with Clamp, Large, 85mm ID, 200mm OAL. Solid ring with built in clamp for attachment directly to support rod. Available in three sizes. Nickel plated zinc alloy.
Catalog No. 50-131-1213 Supplier Chemglass Life Sciences Supplier No. CG305712
Support Ring with Clamp, Large, 85mm ID, 200mm OAL. Solid ring with built in clamp for attachment directly to support rod. Available in three sizes. Nickel plated zinc alloy.