Learn More
Chemglass Life Sciences Support Ring with Clamp, Medium, 64mm ID, 180mm OAL
SDP
Supplier:  Chemglass Life Sciences CG305711
Encompass_Preferred
Support Ring with Clamp, Medium, 64mm ID, 180mm OAL. Solid ring with built in clamp for attachment directly to support rod. Available in three sizes. Nickel plated zinc alloy.
Catalog No. 50-131-1212