Learn More
Med Vet International Support Rings, 3?
Catalog No. 503047219
Encompass_Preferred
Cast iron with clamp to attach to support stand rod. Cast-iron construction prevents breakage under normal use.
Catalog No. 50-304-7219 Supplier Med Vet International Supplier No. 974053
Cast iron with clamp to attach to support stand rod. Cast-iron construction prevents breakage under normal use.