Learn More
Electron Microscopy Sciences Corning Digital Stirring Hot Plate Support Rod, 5/16" x 12"L
SDP
Catalog No. 501899840
Encompass_Preferred
Corning hot plates, stirrers and hot plate/stirrers have a space for a support rod that screws into the base of the unit. The rods are constructed from stainless steel. Size: 5⁄16" x 12"L. Use with models: PC-200/210/220.
Use: PC-200/210/220
Catalog No. 50-189-9840 Supplier Electron Microscopy Sciences Supplier No. 7235112
Corning hot plates, stirrers and hot plate/stirrers have a space for a support rod that screws into the base of the unit. The rods are constructed from stainless steel. Size: 5⁄16" x 12"L. Use with models: PC-200/210/220.
Use: PC-200/210/220