Learn More
Electron Microscopy Sciences Corning Digital Stirring Hot Plate Support Rod, 5/16" x 18"L
SDP
Catalog No. 501899841
Encompass_Preferred
Corning hot plates, stirrers and hot plate/stirrers have a space for a support rod that screws in the base of the unit. The support rods are constructed from stainless steel, size: 5⁄16" x 18"L, use with models: PC-400/410/4/305.
Use: PC-400/410/4/305
Catalog No. 50-189-9841 Supplier Electron Microscopy Sciences Supplier No. 7235118
Corning hot plates, stirrers and hot plate/stirrers have a space for a support rod that screws in the base of the unit. The support rods are constructed from stainless steel, size: 5⁄16" x 18"L, use with models: PC-400/410/4/305.
Use: PC-400/410/4/305