Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Support Rod and Clamp Kit

Catalog No. 021061015 Shop All OHAUS Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-106-1015 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-106-1015 Supplier OHAUS™ Supplier No. 30400146
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OHAUS™ Support Rod and Clamp Kit for holding temperature probes, thermometers, or other glass tubes

  • For all Hotplates or Hotplate-Stirrers
  • Includes Stainless-Steel Support Rod (18 in.), Thermometer/Temperature Probe Extension Clamp, 3-Prong Medium Swivel Clamp and Hook Connector
TRUSTED_SUSTAINABILITY

Specifications

Type Adjustable Clamp
Material Stainless Steel
Includes Support Rod (18 in.), Thermometer/Temperature Probe Extension Clamp, 3-Prong Medium Swivel Clamp and Hook Connector
For Use With (Equipment) Hotplates, Stirrers, and Stirring Hotplates
Description Support Rod And Clamp Kit
Color Silver
Jaw Type Four Interlocking
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.